ENA379364204 Enamine

(2,2-dimethyl-2H-1,3-benzodioxol-5-yl)methanamine

  • MDL number MFCD20726280
  • Cas Number : 1019776-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112907

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-112907
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart