ENA379364205 Enamine

5,7-dimethylquinolin-2-amine

  • MDL number MFCD19678426
  • Cas Number : 1342190-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118285
Availability details for
Quantity
  • Availability
  • Add to Cart