ENA379364209 Enamine

(2-chloro-4,5-dimethylphenyl)methanol

  • MDL number MFCD22578676
  • Cas Number : 1087417-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118290
Availability details for
Quantity
  • Availability
  • Add to Cart