ENA379364218 Enamine

octane-1,3-diol

  • MDL number MFCD01711163
  • Cas Number : 23433-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118308
Availability details for
Quantity
  • Availability
  • Add to Cart