ENA379364221 Enamine

2-[N-(butan-2-yl)3-bromobenzenesulfonamido]acetic acid

  • MDL number MFCD12525662
  • Cas Number : 1183427-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-118312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118312
Availability details for
Quantity
  • Availability
  • Add to Cart