ENA379364225 Enamine

4-[4-(2-methoxyphenyl)-1,3-thiazol-2-yl]morpholine

  • MDL number MFCD03482307
  • Cas Number : 521318-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118323
Availability details for
Quantity
  • Availability
  • Add to Cart