ENA379364244 Enamine

7-chloro-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD09732698
  • Cas Number : 217181-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118403

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118403
Availability details for
Quantity
  • Availability
  • Add to Cart