ENA379364245 Enamine

ethyl 5-amino-2-ethyl-1,3-oxazole-4-carboxylate

  • MDL number MFCD09953576
  • Cas Number : 86186-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118404

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118404
Availability details for
Quantity
  • Availability
  • Add to Cart