ENA379364247 Enamine

6-(tert-butoxy)pyridine-3-carbonitrile

  • MDL number MFCD11196882
  • Cas Number : 435273-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118406
Availability details for
Quantity
  • Availability
  • Add to Cart