ENA379364249 Enamine

decahydronaphthalene-2-carboxylic acid

  • MDL number MFCD00956642
  • Cas Number : 13032-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118408
Availability details for
Quantity
  • Availability
  • Add to Cart