ENA379364257 Enamine

2-chloro-4H,5H,6H-cyclopenta[d][1,3]thiazole

  • MDL number MFCD17249938
  • Cas Number : 201006-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118470
Availability details for
Quantity
  • Availability
  • Add to Cart