ENA379364258 Enamine

1-azabicyclo[3.3.1]nonan-3-one

  • MDL number MFCD22578690
  • Cas Number : 29170-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118480
Availability details for
Quantity
  • Availability
  • Add to Cart