ENA379364260 Enamine

N-(2,2-dimethylpropyl)-4-fluoroaniline

  • MDL number MFCD14625054
  • Cas Number : 1226148-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116639

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116639
Availability details for
Quantity
  • Availability
  • Add to Cart