ENA379364275 Enamine

3-(1,3-dioxolan-2-yl)benzoic acid

  • MDL number MFCD06209118
  • Cas Number : 773101-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118505
Availability details for
Quantity
  • Availability
  • Add to Cart