ENA379364276 Enamine

2-(2,6-difluorophenyl)-2,2-difluoroacetic acid

  • MDL number MFCD19637728
  • Cas Number : 1344264-52-3
  • Molecular Weight : 208.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116646
Availability details for
Quantity
  • Availability
  • Add to Cart