ENA379364280 Enamine

N-methyl-1,3-dioxaindan-5-amine

  • MDL number MFCD11154432
  • Cas Number : 34060-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118510
Availability details for
Quantity
  • Availability
  • Add to Cart