ENA379364281 Enamine

3-hydroxy-3-(4-hydroxyphenyl)-2,2-dimethylpropanoic acid

  • MDL number MFCD16749834
  • Cas Number : 1249919-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116648
Availability details for
Quantity
  • Availability
  • Add to Cart