ENA379364283 Enamine

1,5-diethyl-1H-pyrazol-4-amine

  • MDL number MFCD08700620
  • Cas Number : 1007541-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118514
Availability details for
Quantity
  • Availability
  • Add to Cart