ENA379364284 Enamine

2-methyl-1-(pyrrolidin-2-ylmethyl)-1H-1,3-benzodiazole

  • MDL number MFCD16075762
  • Cas Number : 1249819-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118515
Availability details for
Quantity
  • Availability
  • Add to Cart