ENA379364285 Enamine

2,2-dimethyldodecanoic acid

  • MDL number MFCD00969502
  • Cas Number : 2874-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118517

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118517
Availability details for
Quantity
  • Availability
  • Add to Cart