ENA379364299 Enamine

6-methyl-3,4-dihydro-2H-pyran-5-carboxylic acid

  • MDL number MFCD00179417
  • Cas Number : 5399-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118537
Availability details for
Quantity
  • Availability
  • Add to Cart