ENA379364300 Enamine

2-(oxolan-2-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD21188035
  • Cas Number : 1083369-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118540
Availability details for
Quantity
  • Availability
  • Add to Cart