ENA379364301 Enamine

N-[4-(aminomethyl)phenyl]-2-hydroxybenzamide

  • MDL number MFCD09043589
  • Cas Number : 926271-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118541
Availability details for
Quantity
  • Availability
  • Add to Cart