ENA379364307 Enamine

3-(3-chlorophenyl)-2-methylpropanoic acid

  • MDL number MFCD08445716
  • Cas Number : 66735-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118552

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118552
Availability details for
Quantity
  • Availability
  • Add to Cart