ENA379364308 Enamine

1-[4-(1H-1,2,3-triazol-1-yl)phenyl]ethan-1-amine

  • MDL number MFCD22578709
  • Cas Number : 1423027-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118555
Availability details for
Quantity
  • Availability
  • Add to Cart