ENA379364310 Enamine

3-cyclohexyl-5-phenylimidazolidine-2,4-dione

  • MDL number MFCD12481672
  • Cas Number : 81968-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113121
Availability details for
Quantity
  • Availability
  • Add to Cart