ENA379364311 Enamine

(5S)-2,3,4,5-tetrahydro-1-benzoxepin-5-ol

  • MDL number MFCD21295183
  • Cas Number : 84499-96-7
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-116691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116691
Availability details for
Quantity
  • Availability
  • Add to Cart