ENA379364313 Enamine

2-(2-methylpropyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD09933834
  • Cas Number : 215669-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118582
Availability details for
Quantity
  • Availability
  • Add to Cart