ENA379364314 Enamine

N-[(5-phenyl-1,3,4-oxadiazol-2-yl)methyl]oxan-3-amine

  • MDL number MFCD19137280
  • Cas Number : 1274705-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118584
Availability details for
Quantity
  • Availability
  • Add to Cart