ENA379364319 Enamine

1-(4-methylphenyl)-1-(1H-1,2,4-triazol-1-yl)butan-2-amine

  • MDL number MFCD16286125
  • Cas Number : 1273996-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118627
Availability details for
Quantity
  • Availability
  • Add to Cart