ENA379364325 Enamine

3,5-dichloro-4-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD16112611
  • Cas Number : 24653-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118639

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118639
Availability details for
Quantity
  • Availability
  • Add to Cart