ENA379364326 Enamine

3-amino-2,2-dimethylpropanoic acid hydrochloride

  • MDL number MFCD11858331
  • Cas Number : 2843-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118640
Availability details for
Quantity
  • Availability
  • Add to Cart