ENA379364327 Enamine

1-cyclobutylethan-1-amine

  • MDL number MFCD08704272
  • Cas Number : 60637-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118644
Availability details for
Quantity
  • Availability
  • Add to Cart