ENA379364330 Enamine

8-methyl-3-(piperazin-1-yl)-8-azabicyclo[3.2.1]octane

  • MDL number MFCD12780506
  • Cas Number : 195504-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118649

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118649
Availability details for
Quantity
  • Availability
  • Add to Cart