ENA379364331 Enamine

6-methoxy-2-methylquinazolin-4-ol

  • MDL number MFCD08732971
  • Cas Number : 51413-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265-270
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118651
Availability details for
Quantity
  • Availability
  • Add to Cart