ENA379364338 Enamine

1-ethoxy-2-iodobenzene

  • MDL number MFCD12547815
  • Cas Number : 614-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118658
Availability details for
Quantity
  • Availability
  • Add to Cart