ENA379364352 Enamine

7-oxa-1,3-diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD18333462
  • Cas Number : 81866-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118749

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118749
Availability details for
Quantity
  • Availability
  • Add to Cart