ENA379364355 Enamine

3-[(3-methoxyphenyl)methoxy]-2-methylaniline

  • MDL number MFCD11979642
  • Cas Number : 1154572-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118752
Availability details for
Quantity
  • Availability
  • Add to Cart