ENA379364357 Enamine

3-(2,5-dimethylmorpholin-4-yl)-N'-hydroxy-3-oxopropanimidamide

  • Cas Number : 1158134-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118754
Availability details for
Quantity
  • Availability
  • Add to Cart