ENA379364360 Enamine

3-aminothiane-3-carboxylic acid

  • MDL number MFCD08669346
  • Cas Number : 39124-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118757
Availability details for
Quantity
  • Availability
  • Add to Cart