ENA379364361 Enamine

2-bromo-N-(2-chloro-6-methylphenyl)acetamide

  • MDL number MFCD08844409
  • Cas Number : 918408-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118758
Availability details for
Quantity
  • Availability
  • Add to Cart