ENA379364408 Enamine

2-(chloromethyl)-4-methoxy-6-methylpyridine hydrochloride

  • MDL number MFCD22578727
  • Cas Number : 110374-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118776
Availability details for
Quantity
  • Availability
  • Add to Cart