ENA379364413 Enamine

3-amino-1-cyclopropylpyrrolidin-2-one hydrochloride

  • MDL number MFCD22578732
  • Cas Number : 1423028-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118863
Availability details for
Quantity
  • Availability
  • Add to Cart