ENA379364417 Enamine

2-aminopent-4-ynamide hydrochloride

  • MDL number MFCD22578736
  • Cas Number : 375859-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118938
Availability details for
Quantity
  • Availability
  • Add to Cart