ENA379364421 Enamine

2-amino-N,4-dimethyl-N-(thiolan-3-yl)pentanamide hydrochloride

  • MDL number MFCD22578740
  • Cas Number : 1423028-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118943
Availability details for
Quantity
  • Availability
  • Add to Cart