ENA379364422 Enamine

3-phenyl-5-(pyrrolidin-2-yl)-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD22578741
  • Cas Number : 1423031-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118954
Availability details for
Quantity
  • Availability
  • Add to Cart