ENA379364426 Enamine

7-[4-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD22578745
  • Cas Number : 1423026-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119037
Availability details for
Quantity
  • Availability
  • Add to Cart