ENA379364434 Enamine

4-(chlorosulfonyl)-1-ethyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD22578749
  • Cas Number : 1423031-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43738
Availability details for
Quantity
  • Availability
  • Add to Cart