ENA379364436 Enamine

3-chloro-1-(2-methylpropyl)-1,2-dihydropyrazin-2-one

  • MDL number MFCD22578751
  • Cas Number : 1423025-01-7
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66282
Availability details for
Quantity
  • Availability
  • Add to Cart