ENA379364438 Enamine

{5H,6H,7H,8H-imidazo[1,2-a]pyridin-8-yl}methanamine

  • MDL number MFCD22578754
  • Cas Number : 185796-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94140
Availability details for
Quantity
  • Availability
  • Add to Cart